Rhodium(III) chloride hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:20765-98-4
:
Formula:Cl₃Rh·H₂O
MDL Number:MFCD00149839
:227.28 g/mol
:M-Clarity™ quality level = MQ100
EINECS:233-165-4
Frequently Bought Together