

Iron(III) chloride hexahydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:10025-77-1
:
Formula:FeCl₃·6H₂O
MDL Number:MFCD00149712
:270.3 g/mol
Merck Index:13,04048
Boiling Pt:280…285 °C (1013 hPa)
Melting Pt:37 °C
Density:1.82 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-729-4
UN:3260
ADR:8,III
:GHS05
:GHS07
Frequently Bought Together