Iron(III) chloride hexahydrate 99+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Iron trichloride hexahydrate, Ferric chloride hexahydrate
CAS Number:10025-77-1
:
Formula:FeCl₃·6H₂O
MDL Number:MFCD00149712
Boiling Pt:280…285 °C
Molecular Weight:270.3 g/mol
Melting Pt:37 °C
ADR:8 III
Merck Index:13,04048
EINECS:231-729-4
UN:3260
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Orange Adhering crystalline powder and/or lumps
- Titration Iodometric:99.0 to 102.0 %
- Nitrate (NO3):≤100 ppm
- Sulfate (SO4):≤100 ppm
- Copper (Cu):≤30 ppm
- Zinc (Zn):≤30 ppm
- Ferrous iron (Fe 2+):≤20 ppm
- Phosphorous compounds (as PO4):≤100 ppm
- Insoluble matter:≤0.01 %
Specification Test Results
Frequently Bought Together