Chromium(III) fluoride tetrahydrate, Sigma-Aldrich®
Catalog # SIAL333387-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:181.05 g/mol
ADR:8,II
:M-Clarity™ quality level = MQ100
EINECS:232-137-9
UN:1756
CAS Number:123333-98-2
Formula:CrF₃·4H₂O
MDL Number:MFCD00016045