

Yttrium (III) nitrate hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:13494-98-9
:
Formula:N₃O₉Y·6H₂O
MDL Number:MFCD00149940
:383.01 g/mol
Melting Pt:51.8 °C
Density:2.682 g/cm³ (20 °C)
EINECS:233-802-6
UN:1477
ADR:5.1,III
:M-Clarity™ quality level = MQ200
:GHS03
:GHS07
Frequently Bought Together