

Cadmium(II) nitrate tetrahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10022-68-1
:
Formula:CdN₂O₆·4H₂O
MDL Number:MFCD00149626
:308.48 g/mol
Boiling Pt:132 °C (1013 hPa)
Melting Pt:60 °C
Density:2.5 g/cm³ (20 °C)
EINECS:233-710-6
ADR:5.1,II
:M-Clarity™ quality level = MQ100
UN:3087
:GHS07
:GHS09
Frequently Bought Together