


Copper(II) chloride dihydrate 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10125-13-0
:
Formula:CuCl₂·2H₂O
MDL Number:MFCD00149674
:170.48 g/mol
Merck Index:13,02660
Melting Pt:100 °C (dec.)
Density:2,54 g/cm³ (20 °C)
EINECS:231-210-2
UN:2802
ADR:8,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Blue-green to blue Crystalline powder or needles
- Titration Complexometric:≥97.5 %
Specification Test Results
Frequently Bought Together