Neodymium trinitrate hexahydrate, Sigma-Aldrich®
: Merck
:16454-60-7
:
:N₃NdO₉·6H₂O
:MFCD00149799
:∼74 °C
:2.265 g/cm³ (20 °C)
:5.1,III
:M-Clarity™ quality level = MQ100
:438.35 g/mol
:233-153-9
:1477
:GHS03