
Ammonium heptamolybdate tetrahydrate
Catalog # SERA13370.01
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:1235.86 g/mol
Merck Index:13,00536
Density:2.498 g/cm³ (20 °C)
EINECS:234-722-4
:GHS07
Storage Temperature:Ambient
CAS Number:12054-85-2
:12027-67-7
Formula:(NH₄)₆Mo₇O₂₄·4H₂O
MDL Number:MFCD00167059