
Strontium hydroxide octahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1311-10-0
:
Formula:H₂O₂Sr·8H₂O
MDL Number:MFCD00150395
:265.76 g/mol
Melting Pt:375 °C
Density:1.9 g/cm³ (20 °C)
EINECS:242-367-1
UN:3262
ADR:8,III
:M-Clarity™ quality level = MQ100
:GHS05
Frequently Bought Together