
Strontium hydroxide octahydrate 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1311-10-0
:
Formula:H₂O₂Sr·8H₂O
MDL Number:MFCD00150395
:265.76 g/mol
Melting Pt:100 °C
Density:1,9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:242-367-1
UN:3262
ADR:8,II
:GHS05
Specification Test Results
- Appearance:White Crystalline powder
- Titration with HCl:≥97.5 %
Specification Test Results
Frequently Bought Together