
Calcium nitrate tetrahydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:13477-34-4
:
Formula:Ca(NO₃)₂·4H₂O
MDL Number:MFCD00149604
:236.15 g/mol
Merck Index:13,01687
Melting Pt:∼42 °C
Density:1.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-332-1
UN:1454
ADR:5.1,III
:GHS03
Frequently Bought Together