Magnesium chloride hexahydrate, Sigma-Aldrich®
: Merck
:7791-18-6
:
:MgCl₂·6H₂O
:MFCD00149781
:203.3 g/mol
:13,05684
:1412 °C (1013 hPa)
:117 °C
:1.57 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
:Ambient
:232-094-6
Frequently Bought Together
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals