
Calcium chloride hexahydrate
Catalog # ROTHT886.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
About this item
:
:219.08 g/mol
Merck Index:13,01660
Melting Pt:29.54 °C
Density:1.71 g/cm³ (25 °C)
EINECS:233-140-8
:GHS07
Storage Temperature:Ambient
CAS Number:7774-34-7
Formula:CaCl₂·6H₂O
MDL Number:MFCD00149616