Order Entry
United Kingdom
ContactUsLinkComponent
Tetrasodium diphosphate decahydrate, Sigma-Aldrich®
Irritant
Tetrasodium diphosphate decahydrate, Sigma-Aldrich®
Catalog # SIAL71515-250G
Supplier:  Merck
CAS Number:  
Irritant
Tetrasodium diphosphate decahydrate, Sigma-Aldrich®
Catalog # SIAL71515-250G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Seq:
    3545

Specifications

About this item

 :
 :446.06 g/mol
Melting Pt:990 °C
Density:1.82 g/cm³ (25 °C)
EINECS:231-767-1
 :GHS07
Storage Temperature:Ambient
CAS Number:13472-36-1
Formula:Na₄P₂O₇·10H₂O
MDL Number:MFCD00149200