
di-Sodium tetraborate decahydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:1303-96-4
:
Formula:Na₂B₄O₇·10H₂O
MDL Number:MFCD00149193
:381.37 g/mol
Merck Index:13,08662
Boiling Pt:320 °C (1013 hPa)
Melting Pt:75 °C
Density:1.73 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-540-4
REACH:01-2119490790-32
:GHS08
Frequently Bought Together