tetra-Chloroauric(III) acid trihydrate ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Gold(III) chloride trihydrate, Gold Chloride hydrate, Hydrogen tetrachloroaurate(III) trihydrate, Hydrogen tetrachloroaurate (III) trihydrate, tetra-Chloroauric acid trihydrate
CAS Number:16961-25-4
:
Formula:HAuCl₄·3H₂O
MDL Number:MFCD00149904
Molecular Weight:393.83 g/mol
Melting Pt:30
Storage Temperature:Refrigerator
EINECS:240-948-4
UN:3260
ADR:8 II
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Yellow to orange Powder and/or chunks (May have a brown hue)
- Assay:≥49.0 % (Au)
- Insoluble matter:≤0.1 % (10 % in ether)
- Alkalies+oth.metals (SO4):≤0.2 %
Specification Test Results
Frequently Bought Together