1-Heptanesulphonic acid sodium salt monohydrate, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium 1-heptanesulphonate monohydrate
CAS Number:207300-90-1
:
Formula:C₇H₁₅NaO₃S·1H₂O
MDL Number:MFCD00149550
Molecular Weight:220.27 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:245-210-5
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together