
Oxalic acid dihydrate
Catalog # PROOMM0928.00
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:126.07 g/mol
Merck Index:13,06980
Boiling Pt:149…160 °C (1013 hPa)
Melting Pt:101 °C
Density:1.65 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-634-3
UN:3261
ADR:8,III
REACH:01-2119534576-33
:GHS07
CAS Number:6153-56-6
Formula:HO₂CCO₂H·2H₂O
MDL Number:MFCD00149102