Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Palladium
Palladium 10% on barium sulphate hydrogenation catalyst, Sigma-Aldrich®
Palladium 10% on barium sulphate hydrogenation catalyst, Sigma-Aldrich®
Specifications
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
Environmentally Preferable:
Specifications
About this item
Product Details
:
:
106.420 g/mol
Storage Temperature:
Ambient
EINECS:
231-115-6
CAS Number:
7440-05-3
Formula:
Pd
MDL Number:
MFCD00011167
Product Details