Ammonia solution 30 - 33%
Supplier: Roth Carl
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.88…0.91 g/cm³ (20 °C)
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together