Order Entry
United Kingdom
ContactUsLinkComponent
Ammonia solution 28 - 30%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Ammonia solution 28 - 30%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates
Ammonia solution 28 - 30%, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    -72 °C
  • Boiling Point:
    32 °C
  • Vapor Pressure:
    635 hPa (20 °C)
  • Explosion Limit:
    15.4 - 33.6 %(V)
  • Density:
    0.90 g/cm3 (20 °C)
  • pH:
    (H2O, 20 °C) strongly alkaline
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together