Order Entry
United Kingdom
ContactUsLinkComponent
 
Ammonia solution 28 - 30% (w/w) in water for analysis
 
undefined
Ammonia solution 28 - 30% (w/w) in water for analysis

 

  • Appearance:
    Clear solution
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.002 % (105 °C)
  • Titration with HCl:
    28.0 to 30.0 % (As NH3)
  • Chloride (Cl):
    ≤1 ppm
  • Sulfate (SO4):
    ≤5 ppm
  • Carbonate (as CO2):
    ≤0.006 %
  • Iron (Fe):
    ≤0.25 ppm
  • Heavy metals (as Pb):
    ≤1 ppm
  • Oxidizing substance:
    Passes test

 

 

Frequently Bought Together