Sodium hydroxide 15% in aqueous solution
Supplier: VWR Collection
Certificates
About this item
Synonyms: Caustic soda
CAS Number:1310-73-2
:
Formula:NaOH
MDL Number:MFCD00003548
Boiling Pt:119…135 °C (1013 hPa)
Molecular Weight:40 g/mol
Melting Pt:9…10 °C
ADR:8 II
Merck Index:14,08627
EINECS:215-185-5
UN:1824
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together