

Hydrochloric acid 37% ACS, Fluka™
:7647-01-0
:
:HCl
:MFCD00011324
:42 °C (1.013 hPa)
:–35 °C
:1.190 g/cm³ (20 °C)
:231-595-7
:1789
:8,II
:36.46 g/mol
:GHS07
:GHS05
:Ambient
- Assay:36.5 - 38.0 %
- Residue on ignition (SO₄):Max. 5 ppm
- Arsenic (As):Max. 0.01 ppm
- Iron (Fe):Max. 0.2 ppm
- Vapor Pressure:3.23 psi ( 21.1 °C)
- Vapor Density:1.3 (vs air)
- Heavy metals (ICP) ACS:Max. 1 ppm
- Ammonium (NH₄):Max. 1 ppm
- Bromide (Br):Max. 50 ppm
- Free chlorine (Cl):Max. 1 ppm
- Sulfate (SO₄):Max. 1 ppm
- Sulfite (SO₃):Max. 1 ppm
- APHA:Max. 10
Frequently Bought Together