Order Entry
United Kingdom
ContactUsLinkComponent
Trimethylamine 33% (w/w) in ethanol denatured with 2% cyclohexane
Flammable
Corrosive
Irritant
Trimethylamine 33% (w/w) in ethanol denatured with 2% cyclohexane
Catalog # H27324.AU
CAS Number:  
Flammable
Corrosive
Irritant
Trimethylamine 33% (w/w) in ethanol denatured with 2% cyclohexane
Catalog # H27324.AU
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1.000 mL
  • Seq:
    2865

Specifications

About this item

 :
Merck Index:14,9710
Density:0.75
Flash Pt:–20 °C (–4 °F)
ADR:3,II
 :59.11 g/mol
EINECS:200-875-0
UN:2924
 :GHS02
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:75-50-3
Formula:C₃H₉N
MDL Number:MFCD00008327