Glycolic acid 70%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:79-14-1
:
Formula:C₂H₄O₃
MDL Number:MFCD00004312
:76.05 g/mol
Melting Pt:10 °C
Density:1.27 g/cm³
Storage Temperature:Ambient
EINECS:201-180-5
UN:3265
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Clear yellow to light amber Solution
- Titration with NaOH:65 to 72 % w/w
- GC:≥94 %
- Enantiomeric excess:≥97.5 % (GC)
Specification Test Results
Frequently Bought Together