Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
(±)-1,2-Hexanediol, Sigma-Aldrich®

(±)-1,2-Hexanediol, Sigma-Aldrich®

Supplier:   Merck
(±)-1,2-Hexanediol, Sigma-Aldrich®
Supplier:  Merck
Certificates
(±)-1,2-Hexanediol, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Hexane-1,2-diol, DL-1,2-Hexanediol, (±)-1,2-Dihydroxy hexane
  • CAS Number:6920-22-5
  • Formula:C₆H₁₄O₂
  • MDL Number:MFCD00010737
  • Molecular Weight:118.18 g/mol
  • Boiling Pt:223 °C (1013 hPa)
  • Melting Pt:45 °C
  • Density:0.9507 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ200
  • EINECS:230-029-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - £39.20
    £39.20
    1 * 10 g
       £39.20

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - £96.50
    £96.50
    1 * 50 g
       £96.50

  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - £168.00
    £168.00
    1 * 250 g
       £168.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.