(±)-2-Nonanol
Glass bottle for solids, 1 g
Catalog # ROTH9067.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: (±)-Heptyl methyl carbinol, (±)-2-Hydroxy nonane, (±)-Methyl heptyl carbinol
- CAS Number:628-99-9
- Formula:C₉H₂₀O
- MDL Number:MFCD00004593
- Molecular Weight:144.26 g/mol
- Boiling Pt:193 °C (1013 hPa)
- Melting Pt:–35 °C
- Density:0.831 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:211-065-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



