(±)-2-Amino-1-propanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6168-72-5
:
Formula:C₃H₉NO
MDL Number:MFCD00008085
:75.11 g/mol
Merck Index:12,00488
Boiling Pt:173 °C (1013 hPa)
Melting Pt:8 °C
Density:0.928 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:228-207-3
:GHS05
Storage Temperature:Ambient
Frequently Bought Together