Lactic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:598-82-3
Formula:H₃CCH(OH)COOH
MDL Number:MFCD00004520
Merck Index:13,05351
Boiling Pt:122 °C (20 hPa)
Melting Pt:16.8 °C
Density:1.357 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:113 °C (closed cup)
:90.08 g/mol
EINECS:209-954-4
:GHS05
Storage Temperature:Ambient
Frequently Bought Together