Order Entry
United Kingdom
ContactUsLinkComponent
L(+)-Lactic acid 88.0-92.0%, AnalaR NORMAPUR® analytical reagent
L(+)-Lactic acid 88.0-92.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 101384Q
Supplier:  VWR Chemicals
CAS Number:  
undefined
L(+)-Lactic acid 88.0-92.0%, AnalaR NORMAPUR® analytical reagent
Catalog # 101384Q
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Assay:
    88.0 - 92.0 %
  • IR Spectrum:
    Passes test
  • Reducing substances:
    Passes test
  • Volatile fatty acids:
    Passes test
  • Colouration:
    Max. 20 APHA
  • Relative density:
    1.200 - 1.210
  • Ignition residue (SO4):
    Max. 100 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 50 ppm
  • As (Arsenic):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.02 %
  • Cu (Copper):
    Max. 1 ppm
  • Fe (Iron):
    Max. 3 ppm
  • Pb (Lead):
    Max. 1 ppm
  • Zn (Zinc):
    Max. 10 ppm

Specification Test Results

About this item

 :
 :90.08 g/mol
Merck Index:13,05351
EINECS:200-018-0
 :GHS05
Storage Temperature:Ambient
CAS Number:79-33-4
Formula:C₃H₆O₃
MDL Number:MFCD00064266