Order Entry
United Kingdom
ContactUsLinkComponent
L(+)-Lactic acid 85-90% ACS
L(+)-Lactic acid 85-90% ACS
Catalog # 0349-4L
Supplier:  VWR Chemicals
CAS Number:  
undefined
L(+)-Lactic acid 85-90% ACS
Catalog # 0349-4L
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Chloride:
    <0.001%
  • Heavy Metals (as Pb):
    ≤0.0005%
  • Iron:
    ≤0.0005%
  • Purity:
    85 – 90%
  • Residue on Ignition:
    ≤0.02%
  • Substances Darkened by H2SO4:
    Pass
  • Sulphate:
    ≤0.002%

Specification Test Results

About this item

 :
 :90.08 g/mol
Merck Index:13,05351
Boiling Pt:122 °C (20 hPa)
Melting Pt:16.8 °C
Density:1.357 g/cm³ (20 °C)
Flash Pt:113 °C (closed cup)
Storage Temperature:Ambient
EINECS:209-954-4
 :GHS05
CAS Number:79-33-4
Formula:C₃H₆O₃
MDL Number:MFCD00064266