
3-Bromopropionic acid
Catalog # FLUO003025-100G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:152.98 g/mol
Merck Index:13,01418
Boiling Pt:240 °C (1013 hPa)
Melting Pt:60 °C
Density:1.48 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-694-1
UN:3261
ADR:8,II
:GHS05
CAS Number:590-92-1
Formula:BrH₂CCH₂COOH
MDL Number:MFCD00002763