Order Entry
United Kingdom
ContactUsLinkComponent
 
Polysorbate 20, Sigma-Aldrich®
Catalog # SIAL44112-100G-F
Supplier:  Merck
CAS Number:  
Polysorbate 20, Sigma-Aldrich®
Catalog # SIAL44112-100G-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:500-018-3
CAS Number:9005-64-5
Formula:C₅₈H₁₁₄O₂₆
MDL Number:MFCD00165986