Poly(dimethylsiloxane-co-methylphenylsiloxane), 500 cSt (25°C), Sigma-Aldrich®
Catalog # SIAL378461-50ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:63148-52-7