Order Entry
United Kingdom
ContactUsLinkComponent
Poly(dimethylsiloxane-co-methylphenylsiloxane), 125 cSt (25°C)
Poly(dimethylsiloxane-co-methylphenylsiloxane), 125 cSt (25°C)
  SERA35145.01
 :  Serva
 :  
Poly(dimethylsiloxane-co-methylphenylsiloxane), 125 cSt (25°C)
  SERA35145.01
 :  Serva
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

 

 

 :
 :∼35 °C (1013 hPa)
 :0.982 g/cm³ (25 °C)
 :>101.1 °C (closed cup)
 :Ambient
 :63148-52-7