Bovine serum albumin (BSA)
: Serva
- CAS No.:9048-46-8
90604-29-8 - Source:Serum
- Species:Bovine
- Storage conditions:Refrigerator
- Protein synonyms:BSA|Fraction V|Cohn Fraction V
- Protein/peptide name:Bovine Serum Albumin
- Colour:light yellow
- Grade:SL-5 Grade
Frequently Bought Together