Order Entry
United Kingdom
ContactUsLinkComponent
EMPROVE® Gelatin
EMPROVE® Gelatin
Certificates
EMPROVE® Gelatin

Specifications

  • CAS No.:
    9000-70-8
  • Storage conditions:
    Ambient
  • Form:
    Powder
  • Protein synonyms:
    Gelatine
  • Protein/peptide name:
    Gelatin
  • Grade:
    Ph. Eur., BP, NF

Specifications

Related Information

Frequently Bought Together