

Nickel chromium alloy (80:20 w%) ≥99.9% (metals basis), lumps
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:11106-97-1
:
MDL Number:MFCD00673110
:GHS08
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together