L(+)-Lactic acid ≥90% Ph. Eur.
:79-33-4
:
:C₃H₆O₃
:MFCD00064266
:90.08 g/mol
:122 °C (20 hPa)
:53 °C
:1.11…1.21 g/cm³ (20 °C)
:Ambient
:200-018-0
:GHS05
- Assay:88.0 - 92.0 %
- (S)-enantiomer:≥ 95%
- Appearance:Conforms (see CoA/CoS)
- Identification A:Passes test
- Identification C:Passes test
- Appearance test:Passes test
- Ether-insoluble substances:Passes test
- Sugars and other reducing substances:Passes test
- Citric, oxalic and phosphoric acids:Passes test
- Relative density:1.200 - 1.210
- SO4 (Sulphate):Max. 200 ppm
- Ca (Calcium):Max. 200 ppm
- Heavy metals (as Pb):Max. 10 ppm
- Sulphated ash:Max. 0.1 %
- Residual solvents:Passes test
1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 5L Plastic drum DIN 51 UN (ZPACK13951001)
Packaging Information
20370_LACTIC ACID 90% PH.EUR._LATEX_REV0.pdf
Additional Product Details
spec_VWRC20370.297 spec_VWRC20370.366
Product specification
VWR Chemicals for Biopharma and Pharma Production
Product Literature
Frequently Bought Together