Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Starch (from wheat)

Marketsource
Supplier:   Roth Carl
Starch (from wheat)
Supplier:  Roth Carl
Marketsource
Certificates
Starch (from wheat)

2 Options Available Below

Product Details & Documents
  • CAS Number:9005-25-8
  • Formula:(C₆H₁₀O₅)n
  • MDL Number:MFCD00082026
  • Molecular Weight:162.14 g/mol
  • Density:1.5 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:232-679-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - £29.36
    £29.36
    1 * 1 kg
       £29.36

    Specifications:
    Marketsource
    Marketsource
  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg

    Specifications:
    Marketsource
    Marketsource