Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Agar

Marketsource
Supplier:   Serva
Agar
Supplier:  Serva
Marketsource
Certificates
Agar

2 Options Available Below

Product Details & Documents
Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • MDL Number:MFCD00081288
  • Merck Index:13,00184
  • Melting Pt:90 °C
  • Storage Temperature:Ambient
  • EINECS:232-658-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - £118.64
    £118.64
    1 * 1 kg
       £118.64

    Specifications:
    Marketsource
    Marketsource
  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - £449.43
    £449.43
    1 * 5 kg
       £449.43

    Specifications:
    Marketsource
    Marketsource