Cellulose
Glass bottle for solids, 500 g
Catalog # ROTH5873.1
Marketsource
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Microcrystallinecellulose
- CAS Number:9004-34-6
- Formula:(C₁₂H₂₂O₁₁)n
- MDL Number:MFCD00081512
- Molecular Weight:342.3 g/mol
- Merck Index:13,01977
- Melting Pt:260…270 °C
- Density:1.27…1.6 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:232-674-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



