Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
D-(+)-Glucose, Sigma-Aldrich®

D-(+)-Glucose, Sigma-Aldrich®

Supplier:   Merck
D-(+)-Glucose, Sigma-Aldrich®
Supplier:  Merck
Certificates
D-(+)-Glucose, Sigma-Aldrich®
D-(+)-Glucose, Sigma-Aldrich®

3 Options Available Below

Product Details & Documents
Synonyms:  Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
  • CAS Number:50-99-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148912
  • Molecular Weight:180.16 g/mol
  • Merck Index:14,04459
  • Melting Pt:146 °C
  • Density:1.54 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-075-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 g
    Log in for availability
    Log in for availability
    1 * 250 g
    1 * 250 g - £55.50
    £55.50
    1 * 250 g
       £55.50

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - £189.00
    £189.00
    1 * 1 kg
       £189.00

  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg - £646.00
    £646.00
    1 * 5 kg
       £646.00