Order Entry
United Kingdom
ContactUsLinkComponent
D-(+)-Glucose
D-(+)-Glucose
Catalog # MOLE13002238-1KG
Supplier:  Molekula
CAS Number:  
undefined
D-(+)-Glucose
Catalog # MOLE13002238-1KG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
 :
 :180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912
Melting Pt:146
Density:1.54 g/cm³ (20 °C)
Merck Index:14,04459