Deuterium oxide, Sigma-Aldrich®
Catalog # SIAL343773-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:20 g/mol
Merck Index:12,02984
Boiling Pt:101.3 °C (1013 hPa)
Melting Pt:3.8 °C
Density:1.1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
EINECS:232-148-9
CAS Number:7789-20-0
Formula:D₂O
MDL Number:MFCD00044636