Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Molecular sieve A5 (0.5 nm, 5 Å)
Molecular sieve A5 (0.5 nm, 5 Å)
Molecular sieve A5 (0.5 nm, 5 Å)
Molecular sieve A5 (0.5 nm, 5 Å)
Catalog # ROTH8475.1
Supplier:
Roth Carl
CAS Number:
Expand image
Molecular sieve A5 (0.5 nm, 5 Å)
Catalog # ROTH8475.1
Supplier:
Roth Carl
CAS Number:
Specifications
Pack type:
Glass bottle for solids
Pk:
250 g
Specifications
About this item
Product Details
:
CAS Number:
69912-79-4
:
1318-02-1
Product Details