Cyclooctene
Catalog # ABCRAB140481.0500
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
Specifications
About this item
:
:110.2 g/mol
Boiling Pt:146 °C (1013 hPa)
Melting Pt:–16 °C
Density:0.855 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:213-245-5
UN:3295
ADR:3,III
CAS Number:931-88-4
Formula:C₈H₁₄
MDL Number:MFCD00001753