Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rhodium
Rhodium ≥99.95% (metals basis), slug
Rhodium ≥99.95% (metals basis), slug
Rhodium ≥99.95% (metals basis), slug
13381.KG
:
Thermo Fisher Scientific
:
Rhodium ≥99.95% (metals basis), slug
13381.KG
:
Thermo Fisher Scientific
:
Pk:
1
Seq:
0001
:
:
12,08353
:
3400…3700 °C (1013 hPa)
:
1960 °C
:
12.41 g/cm³ (20 °C)
:
102.9 g/mol
:
231-125-0
:
7440-16-6
:
Rh
:
MFCD00011201